4,6-Dichloro-2-(difluoromethyl)pyrimidine 95%

Supplier: Ambeed
Warning

A101821-100MG A101821-250MG A101821-1G
103796-158EA 150 USD
103796-158 103821-848 77257-254 77257-256 77308-742
4,6-Dichloro-2-(difluoromethyl)pyrimidine 95%
4,6-Dichloro-2-(difluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂F₂N₂
MW: 198.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29045042
CAS Number: 1816289-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR