4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine 97%

Supplier: Ambeed
Warning

A136913-10G A136913-250MG A136913-1G A136913-25G A136913-5G
103802-372EA 33 USD
103802-372 76782-704 76782-706 77298-872 77315-294 77315-296
4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine 97%
4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₄
Boiling Pt: 283.9 °C (760 mm Hg)
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750213
CAS Number: 98141-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR