4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

A137639-100MG A137639-10G A137639-1G A137639-5G A137639-250MG
103802-480EA 103 USD
103802-480 77285-608 77285-610 77315-452 77315-454 77747-974
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine 97%
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
Boiling Pt: 371.6 °C (760 mm Hg)
Flash Pt: 210.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038480
CAS Number: 67139-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR