4,6-Dichloro-7-azaindole 95%

Supplier: Ambeed
Warning

A319560-10G A319560-1G A319560-25G A319560-250MG
103825-206EA 23 USD
103825-206 103836-138 76788-046 76838-986 76838-988 77668-418
4,6-Dichloro-7-azaindole 95%
4,6-Dichloro-7-azaindole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
Boiling Pt: 319.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09880157
CAS Number: 5912-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR