4,6-Dichloro-1H-pyrazolo[3,4-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A174726-100MG
A174726-250MG
103806-970EA
14
USD
103806-970
103845-270
77286-974
77286-976
4,6-Dichloro-1H-pyrazolo[3,4-d]pyrimidine 97%
4,6-Dichloro-1H-pyrazolo[3,4-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₅H₂Cl₂N₄ MW: 189.0022 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09835493 CAS Number: 42754-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...