4,6-Dichloroindole 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloro-1H-indole

A157075-250MG A157075-1G
103804-958EA 42 USD
103804-958 103843-160 77110-532 77318-974
4,6-Dichloroindole 97%
4,6-Dichloroindole

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Boiling Pt: 328.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03094990
CAS Number: 101495-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR