4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A416260-100MG A416260-250MG
103827-308EA 68 USD
103827-308 103838-354 76990-398 76990-400
4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one 95%
4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃O
MW: 204.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528104
CAS Number: 668268-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR