4,6-Dichloro-1,7-naphthyridine 98%

Supplier: Ambeed
Warning

A570127-100MG A570127-250MG A570127-50MG A570127-1G
103828-800EA 1013 USD
103828-800 77157-310 77157-312 77157-314 77307-850
4,6-Dichloro-1,7-naphthyridine 98%
4,6-Dichloro-1,7-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18837777
CAS Number: 1301714-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR