4,6-Dibromothieno[3,4-b]thiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A212287-100MG A212287-250MG A212287-1G
103818-184EA 419 USD
103818-184 76986-764 76986-766 77087-348
4,6-Dibromothieno[3,4-b]thiophene-2-carboxylic acid 97%
4,6-Dibromothieno[3,4-b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₂Br₂O₂S₂
MW: 342.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19440952
CAS Number: 1024594-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR