4,6-Dibromopyridazin-3-amine 95%

Supplier: Ambeed
Warning

A317947-100MG A317947-250MG A317947-1G A317947-5G
103825-156EA 70 USD
103825-156 103836-086 76828-128 76828-130 76838-934 76838-936
4,6-Dibromopyridazin-3-amine 95%
4,6-Dibromopyridazin-3-amine

Precaución: for Research Use Only

Formula: C₄H₃Br₂N₃
Boiling Pt: 376.6±37.0 °C (760 mm Hg)
Flash Pt: 181.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13689240
CAS Number: 1206487-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR