4,6-Dibromo-1H-indole 95%

Supplier: Ambeed
Warning

A294813-100MG A294813-250MG A294813-1G
103814-030EA 183 USD
103814-030 77289-850 77289-852 77305-664
4,6-Dibromo-1H-indole 95%
4,6-Dibromo-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
Boiling Pt: 368.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07357291
CAS Number: 99910-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR