4,6-Bis(diphenylphosphino)phenoxazine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Bis(diphenylphosphino)-10H-phenoxazine, NIXANTPHOS

A122157-250MG A122157-25G A122157-1G A122157-5G A122157-10G
103800-182EA 12 USD
103800-182 77284-858 77284-860 77312-318 77312-320 77566-298
4,6-Bis(diphenylphosphino)phenoxazine 97%
4,6-Bis(diphenylphosphino)phenoxazine

Precaución: For research use only.

Formula: C₃₆H₂₇NOP₂
Boiling Pt: 662.1±55.0 °C (760 mm Hg)
Melting Pt: 256…262 °C
Flash Pt: 354.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03788937
CAS Number: 261733-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR