Pyrroloquinoline quinone 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dioxo-4,5-dihydro-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, PQQ, Methoxatin

A309883-100G A309883-100MG A309883-250MG A309883-1G
103814-604EA 63 USD
103814-604 103835-784 76802-734 76802-736 76802-738 76838-560
Pyrroloquinoline quinone 97%
Pyrroloquinoline quinone

Precaución: for Research Use Only

Formula: C₁₄H₆N₂O₈
MW: 330.21 g/mol
Boiling Pt: 1018.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00043125
CAS Number: 72909-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR