4,5-Dimethylpyridazine-3,6-diol 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dihydroxy-4,5-dimethyl-1,2-diazine, 3,6-Dihydroxy-4,5-dimethylpyridazine

A147247-10G A147247-25G A147247-5G
103803-754EA 6 USD
103803-754 103841-852 77285-990 77285-992 77317-220
4,5-Dimethylpyridazine-3,6-diol 97%
4,5-Dimethylpyridazine-3,6-diol

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 422.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00067744
CAS Number: 5754-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR