4,5-Dimethyl-o-phenylenediamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene
A310968-25G
A310968-5G
A310968-100G
A310968-10G
A310968-500G
A310968-1G
103814-646EA
5
USD
103814-646
103835-832
76838-612
76838-614
77194-628
77194-630
77194-632
77306-042
4,5-Dimethyl-o-phenylenediamine 98%
4,5-Dimethyl-o-phenylenediamine
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂ Boiling Pt: 279.6 °C (760 mm Hg) Melting Pt: 127…129 °C Flash Pt: 144.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007729 CAS Number: 3171-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...