2-Nitro-4,5-xylidine 98%

Supplier: Ambeed
Warning

A701738-5G A701738-100G A701738-10G A701738-1G A701738-25G
103840-646EA 10 USD
103840-646 76844-864 77198-836 77198-838 77198-840 77308-162
2-Nitro-4,5-xylidine 98%
2-Nitro-4,5-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 335.7 °C (760 mm Hg)
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00007811
CAS Number: 6972-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR