4,5-Diiodoimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Diiodo-1H-imidazole

A623241-1G A623241-25G A623241-100G A623241-10G A623241-250MG
103840-228EA 20 USD
103840-228 76844-396 76844-398 77005-524 77005-526 77005-528
4,5-Diiodoimidazole 97%
4,5-Diiodoimidazole

Precaución: for Research Use Only

Formula: C₃H₂I₂N₂
MW: 319.87 g/mol
Boiling Pt: 436.9 °C (760 mm Hg)
Melting Pt: 197…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00963878
CAS Number: 15813-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR