4,5-Difluoropyridin-2-amine 95%

Supplier: Ambeed
Warning

A285279-100MG A285279-250MG A285279-50MG
103813-662EA 480 USD
103813-662 77014-648 77612-640 77683-856
4,5-Difluoropyridin-2-amine 95%
4,5-Difluoropyridin-2-amine

Precaución: For research use only.

Formula: C₅H₄F₂N₂
MW: 130.1 g/mol
Boiling Pt: 205.7±35.0 °C (760 mm Hg)
Flash Pt: 78.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18259315
CAS Number: 1211537-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR