4,5-Difluorophthalic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluorophthalic Anhydride

A231148-100MG A231148-10G A231148-250MG A231148-1G A231148-5G A231148-25G
103810-756EA 12 USD
103810-756 103850-504 76771-152 76771-154 76771-156 77303-902 77303-904 77593-496
4,5-Difluorophthalic anhydride 98%
4,5-Difluorophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₂F₂O₃
MW: 184.1 g/mol
Boiling Pt: 321.2 °C (760 mm Hg)
Flash Pt: 142.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08695033
CAS Number: 18959-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR