4,5-Difluorophthalic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluorobenzene-1,2-dicarboxylic acid

A165077-10G A165077-250MG A165077-1G A165077-5G
103805-942EA 8 USD
103805-942 103844-200 76768-774 76768-776 77320-396 77320-398
4,5-Difluorophthalic acid 97%
4,5-Difluorophthalic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₄
MW: 202.112 g/mol
Boiling Pt: 378.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00077514
CAS Number: 18959-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR