4,5-Difluoro-o-toluidine 95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-2-methylaniline

A532681-1G A532681-5G A532681-10G A532681-250MG
103828-582EA 18 USD
103828-582 103839-694 76843-708 76843-710 77292-330 77292-332
4,5-Difluoro-o-toluidine 95%
4,5-Difluoro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₇F₂N
Boiling Pt: 216.2 °C (760 mm Hg)
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08062378
CAS Number: 875664-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR