4,5-Dichloropyridin-3-amine 97%

Supplier: Ambeed
Warning

A260054-100MG A260054-250MG
103812-476EA 68 USD
103812-476 103852-358 77276-532 77276-534
4,5-Dichloropyridin-3-amine 97%
4,5-Dichloropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09910182
CAS Number: 89284-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR