2-Amino-4,5-dichloropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dichloropyridin-2-amine

A135080-250MG A135080-1G A135080-5G
103802-088EA 19 USD
103802-088 77109-474 77314-872 77314-874
2-Amino-4,5-dichloropyridine 97%
2-Amino-4,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.0100 g/mol
Boiling Pt: 269.8 °C (760 mm Hg)
Flash Pt: 116.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07437842
CAS Number: 188577-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR