4,5-Dichloropicolinic acid 95%

Supplier: Ambeed
Warning

A193710-100MG A193710-250MG
103799-460EA 58 USD
103799-460 103816-570 76974-940 76974-942
4,5-Dichloropicolinic acid 95%
4,5-Dichloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 191.99 g/mol
Boiling Pt: 328.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11100716
CAS Number: 73455-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR