4,5-Dichloro-o-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine

A141660-100G A141660-10G A141660-1G A141660-25G A141660-5G
103803-034EA 5 USD
103803-034 77285-772 77285-774 77299-234 77299-236 77316-204
4,5-Dichloro-o-phenylenediamine 98%
4,5-Dichloro-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 346.3 °C (760 mm Hg)
Melting Pt: 158…164 °C
Flash Pt: 163.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007723
CAS Number: 5348-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR