4,5-Dichloro-6-methylpyrimidine 95%

Supplier: Ambeed
Warning

A299552-250MG A299552-5G
103814-230EA 45 USD
103814-230 77668-330 77872-397
4,5-Dichloro-6-methylpyrimidine 95%
4,5-Dichloro-6-methylpyrimidine

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₅H₄Cl₂N₂
Boiling Pt: 242.6±35.0 °C (760 mm Hg)
Flash Pt: 123.983 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909763
CAS Number: 83942-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR