4,5-Dichloro-2-nitroaniline 95%

Supplier: Ambeed
Warning

A158086-100G A158086-500G A158086-5G A158086-25G A158086-10G
103843-300EA 6 USD
103843-300 77096-074 77096-076 77300-312 77319-144 77900-615
4,5-Dichloro-2-nitroaniline 95%
4,5-Dichloro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
Melting Pt: 177…179 °C
Flash Pt: 163.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007770
CAS Number: 6641-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR