4,5-Dichloro-2-methyl-2H-pyridazin-3-one 98%
Supplier: Ambeed
|
Danger
|
A357393-1G
A357393-25G
A357393-5G
A357393-10G
A357393-250MG
103826-190EA
6
USD
103826-190
103837-194
76840-344
76840-346
76840-348
77290-714
77290-716
4,5-Dichloro-2-methyl-2H-pyridazin-3-one 98%
4,5-Dichloro-2-methyl-2H-pyridazin-3-one
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂O Boiling Pt: 197.2 °C (760 mm Hg) Melting Pt: 87…91 °C Flash Pt: 73.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051686 CAS Number: 933-76-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...