4,5-Dibromo-3(2H)-pyridazinone 95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dibromopyridazin-3(2H)-one

A149942-100G A149942-10G A149942-500G A149942-1G A149942-25G A149942-5G
103804-106EA 7 USD
103804-106 103842-234 77110-246 77110-248 77110-250 77299-764 77317-750 77317-752
4,5-Dibromo-3(2H)-pyridazinone 95%
4,5-Dibromo-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₄H₂Br₂N₂O
MW: 253.89 g/mol
Melting Pt: 231…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00023641
CAS Number: 5788-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR