4,5-Dibromo-2-furoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,5-Dibromofuran-2-carboxylic acid
A274502-100G
A274502-10G
A274502-250MG
A274502-1G
A274502-25G
103813-198EA
14
USD
103813-198
103853-126
76988-350
76988-352
76988-354
77092-946
77092-948
4,5-Dibromo-2-furoic acid 97%
4,5-Dibromo-2-furoic acid
Precaución: for Research Use Only
Formula:
C₅H₂Br₂O₃ Boiling Pt: 400.2 °C (760 mm Hg) Melting Pt: 222…224 °C Flash Pt: 152.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092311 CAS Number: 2434-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...