4,5-Dibromo-2H-1,2,3-triazole 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A190726-100G
A190726-10G
A190726-250MG
A190726-1G
A190726-25G
A190726-5G
A190726-500G
103816-324EA
6
USD
103816-324
103846-926
76986-098
76986-100
76986-102
77084-746
77084-748
77084-750
77683-499
4,5-Dibromo-2H-1,2,3-triazole 98%, Ambeed.Inc
4,5-Dibromo-2H-1,2,3-triazole
Precaución: For research use only.
Formula:
C₂HBr₂N₃ Boiling Pt: 347.5±22.0 °C (760 mm Hg) Flash Pt: 163.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD19440717 CAS Number: 22300-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...