4,5-Dibromo-1H-1,2,3-triazole 98%

Supplier: Ambeed
Warning

A109147-100G A109147-10G A109147-250MG A109147-25G A109147-1G A109147-5G A109147-500G
103797-454EA 6 USD
103797-454 103823-246 77283-812 77283-814 77283-816 77283-818 77310-062 77310-064 77747-806
4,5-Dibromo-1H-1,2,3-triazole 98%
4,5-Dibromo-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₂HBr₂N₃
Boiling Pt: 347.5 °C (760 mm Hg)
Melting Pt: 195…199 °C
Flash Pt: 163.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062401
CAS Number: 15294-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR