4,5-Diaminophthalonitrile 96%

Supplier: Ambeed
Danger

A118573-100MG A118573-10G A118573-250MG A118573-5G A118573-25G
103799-076EA 33 USD
103799-076 103799-304 76810-332 76810-334 76810-336 77311-744 77610-976
4,5-Diaminophthalonitrile 96%
4,5-Diaminophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₄
Melting Pt: 270…276 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02376082
CAS Number: 129365-93-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR