4,5-Bis(dicyclohexylphosphino)-9,9-dimethyl-9H-xanthene 97%

Supplier: Ambeed
Warning

A207502-25G A207502-1G A207502-5G A207502-250MG
103817-800EA 25 USD
103817-800 103848-482 77030-362 77086-758 77086-760 77667-742
4,5-Bis(dicyclohexylphosphino)-9,9-dimethyl-9H-xanthene 97%
4,5-Bis(dicyclohexylphosphino)-9,9-dimethyl-9H-xanthene

Precaución: for Research Use Only

Formula: C₃₉H₅₆OP₂
MW: 602.81 g/mol
Boiling Pt: 390.9 °C (760 mm Hg)
Melting Pt: 94…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11559064
CAS Number: 940934-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR