4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-2-carboxylic acid

A238949-100MG A238949-250MG A238949-1G
103811-256EA 116 USD
103811-256 103851-052 76722-394 76722-396 77304-126
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid 97%
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.177 g/mol
Boiling Pt: 397.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09056814
CAS Number: 307313-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR