4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A182772-250MG A182772-1G A182772-100MG
103815-564EA 205 USD
103815-564 103846-120 77012-048 77083-678 77683-460
4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine 95%, Ambeed.Inc
4,5,6,7-Tetrahydro-2-methylthiazolo[5,4-c]pyridine

Precaución: For research use only.

Formula: C₇H₁₀N₂S
MW: 154.2327 g/mol
Boiling Pt: 273.3 °C (760 mm Hg)
Flash Pt: 119.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738531
CAS Number: 124458-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR