Bis(4-fluorophenyl)ether 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Oxybis(fluorobenzene), 4,4'-Difluorodiphenyl Ether

A298105-250MG A298105-1G
103814-172EA 13 USD
103814-172 103835-316 76772-556 76838-014
Bis(4-fluorophenyl)ether 95%
Bis(4-fluorophenyl)ether

Precaución: for Research Use Only

Formula: C₁₂H₈F₂O
MW: 206.19 g/mol
Boiling Pt: 248 °C (760 mm Hg)
Flash Pt: 110.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00013552
CAS Number: 330-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR