4,4'-Dihydroxydiphenylmethane 98%
Supplier: Ambeed
|
Warning
|
A222523-1G
A222523-25G
A222523-5G
A222523-100G
A222523-250MG
103818-980EA
6
USD
103818-980
103849-862
77088-466
77088-468
77088-470
77288-248
77288-250
4,4'-Dihydroxydiphenylmethane 98%
4,4'-Dihydroxydiphenylmethane
Precaución: for Research Use Only
Formula:
C₁₃H₁₂O₂ Melting Pt: 162…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002385 CAS Number: 620-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...