4,4'-Di-tert-butyl-2,2'-bipyridine 98%

Supplier: Ambeed
Warning

A105158-100G A105158-10G A105158-250MG A105158-25G A105158-1G A105158-5G
103796-850EA 9 USD
103796-850 103822-578 76809-540 76809-542 76809-544 76809-546 77309-450 77309-452
4,4'-Di-tert-butyl-2,2'-bipyridine 98%
4,4'-Di-tert-butyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂
MW: 268.4000 g/mol
Boiling Pt: 395.4 °C (760 mm Hg)
Melting Pt: 159…161 °C
Flash Pt: 150.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01863731
CAS Number: 72914-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR