4,4'-Di-tert-butylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl

A322793-100G A322793-10G A322793-25G A322793-5G
103836-246EA 6 USD
103836-246 77290-324 77290-326 77306-274 77306-276
4,4'-Di-tert-butylbiphenyl 97%
4,4'-Di-tert-butylbiphenyl

Precaución: for Research Use Only

Formula: C₂₀H₂₆
MW: 266.43 g/mol
Boiling Pt: 228
Melting Pt: 127
Storage Temperature: Ambient
MDL Number: MFCD00008834
CAS Number: 1625-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR