4,4'-Dipyridyl disulfide 98%

Supplier: Ambeed
Warning

Synonyms: 4,4-Dipyridyl Disulphide, 4,4'-Dithiodipyridine

A398614-1G A398614-5G A398614-10G A398614-250MG
103827-020EA 20 USD
103827-020 103838-060 76841-492 76841-494 77291-120 77291-122
4,4'-Dipyridyl disulfide 98%
4,4'-Dipyridyl disulphide

Precaución: for Research Use Only

Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 356.1 °C (760 mm Hg)
Melting Pt: 75…79 °C at N/A mmHg
Flash Pt: 169.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006423
CAS Number: 2645-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR