4,4-Dimethyl-2-pyrrolidinone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4-Dimethylpyrrolidin-2-one
A183996-250MG
A183996-25G
103815-696EA
77
USD
103815-696
103846-256
77287-356
77301-870
4,4-Dimethyl-2-pyrrolidinone 97%
4,4-Dimethyl-2-pyrrolidinone
Precaución: for Research Use Only
Formula:
C₆H₁₁NO Melting Pt: 38…40 °C Flash Pt: 121.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864496 CAS Number: 66899-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...