4,4-Dimethylpiperidine hydrochloride 97%

Supplier: Ambeed
Warning

A111949-100G A111949-10G A111949-250MG A111949-25G A111949-1G A111949-5G
103797-924EA 18 USD
103797-924 103823-748 76995-256 76995-258 76995-260 76995-262 77310-554 77310-556
4,4-Dimethylpiperidine hydrochloride 97%
4,4-Dimethylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClN
Boiling Pt: 181.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00035295
CAS Number: 38646-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR