4,4'-Dimethyldiphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: p,p'-Ditolylamine, 4,4'-Dimethyldiphenylamine

A173472-10G A173472-500G A173472-25G A173472-5G A173472-100G
103845-116EA 9 USD
103845-116 76784-314 76784-316 77301-268 77301-270 77600-408
4,4'-Dimethyldiphenylamine 98%
4,4'-Dimethyldiphenylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
Boiling Pt: 330 °C (760 mm Hg)
Melting Pt: 78…82 °C
Flash Pt: 167.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00059317
CAS Number: 620-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR