4,4-Dimethylcyclohexanone 98%

Supplier: Ambeed
Danger

A316424-1G A316424-100G A316424-10G A316424-25G A316424-5G
103825-100EA 7 USD
103825-100 103836-026 76838-868 77290-222 77290-224 77290-226 77306-146 77306-148
4,4-Dimethylcyclohexanone 98%
4,4-Dimethylcyclohexanone

Precaución: for Research Use Only

Formula: C₈H₁₄O
Boiling Pt: 170.6 °C (760 mm Hg)
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00234965
CAS Number: 4255-62-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR