4,4-Dimethylcyclohexanol 97%

Supplier: Ambeed
Warning

A519007-5G A519007-10G A519007-250MG A519007-1G
103828-486EA 38 USD
103828-486 103839-594 76843-598 77292-262 77292-264 77307-704
4,4-Dimethylcyclohexanol 97%
4,4-Dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₈H₁₆O
MW: 128.2100 g/mol
Melting Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00101954
CAS Number: 932-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR