4,4-Dimethylcyclohexanamine 97%
Supplier: Ambeed
|
Danger
|
A421546-100MG
A421546-10G
A421546-250MG
A421546-1G
A421546-5G
103827-406EA
172
USD
103827-406
103838-452
77291-424
77291-426
77291-428
77307-216
77307-218
4,4-Dimethylcyclohexanamine 97%
4,4-Dimethylcyclohexanamine
Precaución: Keep in a dark place, inert atmosphere, room temperature
Formula:
C₈H₁₇N Boiling Pt: 160.5 °C (760 mm Hg) Flash Pt: 30.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD08460046 CAS Number: 20615-18-3 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...