4,4'-Dimethylbenzophenone 97%

Supplier: Ambeed
Warning

A112054-100G A112054-10G A112054-25G A112054-500G A112054-5G
103823-772EA 5 USD
103823-772 76781-158 76781-160 76781-162 76781-164 77310-578
4,4'-Dimethylbenzophenone 97%
4,4'-Dimethylbenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Boiling Pt: 354.1 °C (760 mm Hg)
Melting Pt: 93…97 °C
Flash Pt: 153.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00017214
CAS Number: 611-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR