4,4'-Dimethyl-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

A170802-25G A170802-5G A170802-100G A170802-10G A170802-500G A170802-1G
103806-544EA 9 USD
103806-544 103844-820 77081-938 77081-940 77273-716 77273-718 77273-720 77301-078
4,4'-Dimethyl-2,2'-bipyridyl 98%
4,4'-Dimethyl-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 169…174 °C
Flash Pt: 116.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00006441
CAS Number: 1134-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR