4,4'-Dimethoxytrityl chloride 98%
Supplier: Ambeed
|
Danger
|
A349428-100G
A349428-10G
A349428-500G
A349428-25G
A349428-5G
103810-192EA
271
USD
103810-192
103837-010
76803-260
76803-262
76803-264
76803-266
76840-102
77306-600
77306-602
4,4'-Dimethoxytrityl chloride 98%
4,4'-Dimethoxytrityl chloride
Precaución: for Research Use Only
Formula:
C₂₁H₁₉ClO₂ Boiling Pt: 463.1 °C (760 mm Hg) Melting Pt: 123…125 °C Flash Pt: 155.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008409 CAS Number: 40615-36-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...